Assay Details
| Gene Symbol: SOX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077645.1 | XP_020933304.1 | 1142 | 87 | ||
| GenBank mRNA | AB271934.1 | - | 2 | 751 | 87 | |
| AK394874.1 | - | 2 | 1132 | 87 | ||
More Information
Set Membership: |
|

SOX18
Genomic Map