Assay Details
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003358030.4 | XP_003358078.1 | 2182 | 63 | ||
XM_005656956.3 | XP_005657013.1 | 5563 | 63 | |||
XM_005656957.3 | XP_005657014.1 | 5497 | 63 | |||
XM_005656958.3 | XP_005657015.1 | 5497 | 63 | |||
XM_013989935.2 | XP_013845389.1 | 5500 | 63 | |||
XM_021066924.1 | XP_020922583.1 | 5420 | 63 | |||
XM_021066925.1 | XP_020922584.1 | 5560 | 63 | |||
XM_021066926.1 | XP_020922585.1 | 6206 | 63 | |||
XM_021066927.1 | XP_020922586.1 | 5228 | 63 | |||
XM_021066928.1 | XP_020922587.1 | 5492 | 63 | |||
XM_021066929.1 | XP_020922588.1 | 5465 | 63 | |||
XM_021066930.1 | XP_020922589.1 | 2959 | 63 | |||
XM_021066931.1 | XP_020922590.1 | 3848 | 63 | |||
XM_021066932.1 | XP_020922591.1 | 2893 | 63 | |||
XM_021066933.1 | XP_020922592.1 | 2890 | 63 | |||
XM_021066934.1 | XP_020922593.1 | 2248 | 63 | |||
XM_021066935.1 | XP_020922594.1 | 2182 | 63 | |||
XM_021066936.1 | XP_020922595.1 | 2179 | 63 | |||
GenBank mRNA | AB271921.1 | - | 18 - 19 | 5260 | 63 | |
AB355637.1 | - | 10 - 11 | 4449 | 63 | ||
AK348349.1 | - | 9 - 10 | 1160 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
