Assay Details
| Gene Symbol: OPRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355059.5 | XP_003355107.2 | 1241 | 78 | ||
| GenBank mRNA | AF012105.1 | - | 2 | 654 | 78 | |
| L49437.1 | - | 2 | 675 | 78 | ||
More Information
Set Membership: |
|

OPRK1
Genomic Map