Assay Details
| Gene Symbol: RGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357617.4 | XP_003357665.1 | 511 | 86 | ||
| XM_005667913.3 | XP_005667970.1 | 472 | 86 | |||
| GenBank mRNA | AF139837.1 | - | 1 | 54 | 86 | |
More Information
Set Membership: |
|

RGS1
Genomic Map