Assay Details
| Gene Symbol: TIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145985.1 | NP_001139457.1 | 2 | 192 | 77 | |
| XM_013989846.2 | XP_013845300.1 | 79 | 77 | |||
| XM_021066201.1 | XP_020921860.1 | 262 | 77 | |||
| XM_021066202.1 | XP_020921861.1 | 178 | 77 | |||
| GenBank mRNA | AF156030.1 | - | 1 | 49 | 77 | |
| FJ394384.1 | - | 2 | 192 | 77 | ||
More Information
Set Membership: |
|

TIMP2
Genomic Map