Assay Details
| Gene Symbol: BGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135475.5 | XP_003135523.1 | 1191 | 74 | ||
| XM_013986528.2 | XP_013841982.2 | 1157 | 74 | |||
| GenBank mRNA | AF159382.1 | - | 3 | 254 | 74 | |
| AK396411.1 | - | 7 | 1010 | 74 | ||
More Information
Set Membership: |
|

BGN
Genomic Map