Assay Details
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980908.2 | XP_013836362.1 | 306 | 67 | ||
| GenBank mRNA | AF203915.1 | - | 1 | 46 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980908.2 | XP_013836362.1 | 306 | 67 | ||
| GenBank mRNA | AF203915.1 | - | 1 | 46 | 67 | |
Set Membership: |
|