Assay Details
| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067989.1 | XP_020923648.1 | 443 | 130 | ||
| XM_021067990.1 | XP_020923649.1 | 596 | 130 | |||
| XM_021067992.1 | XP_020923651.1 | 883 | 130 | |||
| XM_021067993.1 | XP_020923652.1 | 491 | 130 | |||
| GenBank mRNA | AF239165.1 | - | 2 | 143 | 130 | |
| AK345948.1 | - | 4 | 353 | 130 | ||
| AK346004.1 | - | 4 | 369 | 130 | ||
| AK346138.1 | - | 4 | 802 | 130 | ||
| AK393991.1 | - | 4 | 530 | 130 | ||
| AK394109.1 | - | 4 | 348 | 130 | ||
More Information
Set Membership: |
|

SHMT1
Genomic Map