Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244430.1 | NP_001231359.1 | 2 | 809 | 138 | |
| GenBank mRNA | AF288820.1 | - | 1 | 152 | 138 | |
| AK234723.1 | - | 2 | 760 | 138 | ||
| AK395310.1 | - | 3 | 521 | 138 | ||
| AK397432.1 | - | 2 | 772 | 138 | ||
| AY609752.1 | - | 2 | 724 | 138 | ||
More Information
Set Membership: |
|

DNAJB1
Genomic Map