Assay Details
| Gene Symbol: EIF4EBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244225.1 | NP_001231154.1 | 2 | 212 | 71 | |
| GenBank mRNA | AF337389.1 | - | 2 | 148 | 71 | |
| AK399690.1 | - | 2 | 239 | 71 | ||
| AY610291.1 | - | 2 | 212 | 71 | ||
More Information
Set Membership: |
|

EIF4EBP1
Genomic Map