Assay Details
| Gene Symbol: PLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086465.1 | XP_020942124.1 | 2344 | 131 | ||
| GenBank mRNA | AF339021.1 | - | 5 | 752 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086465.1 | XP_020942124.1 | 2344 | 131 | ||
| GenBank mRNA | AF339021.1 | - | 5 | 752 | 131 | |
Set Membership: |
|