Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134092.6 | XP_003134140.2 | 570 | 98 | ||
| XM_005672563.3 | XP_005672620.1 | 578 | 98 | |||
| XM_005672564.3 | XP_005672621.1 | 557 | 98 | |||
| XM_005672565.3 | XP_005672622.1 | 567 | 98 | |||
| XM_005672566.3 | XP_005672623.1 | 575 | 98 | |||
| XM_013984882.2 | XP_013840336.1 | 520 | 98 | |||
| XM_021076846.1 | XP_020932505.1 | 556 | 98 | |||
| GenBank mRNA | AF339886.1 | - | 3 | 251 | 98 | |
| AK344351.1 | - | 3 | 2407 | 98 | ||
| AY609542.1 | - | 5 | 490 | 98 | ||
More Information
Set Membership: |
|

PTTG1
Genomic Map