Assay Details
| Gene Symbol: FGF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013993026.2 | XP_013848480.2 | 707 | 77 | ||
| XM_021073771.1 | XP_020929430.1 | 532 | 77 | |||
| GenBank mRNA | AJ245447.1 | - | 1 | 48 | 77 | |
More Information
Set Membership: |
|

FGF7
Genomic Map