Assay Details
| Gene Symbol: PCDH11X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673778.3 | XP_005673835.1 | 288 | 67 | ||
| XM_021080409.1 | XP_020936068.1 | 725 | 67 | |||
| XM_021080410.1 | XP_020936069.1 | 288 | 67 | |||
| XM_021080411.1 | XP_020936070.1 | 697 | 67 | |||
| XM_021080413.1 | XP_020936072.1 | 725 | 67 | |||
| XM_021080414.1 | XP_020936073.1 | 725 | 67 | |||
| XM_021080415.1 | XP_020936074.1 | 725 | 67 | |||
| XM_021080416.1 | XP_020936075.1 | 725 | 67 | |||
| XM_021080417.1 | XP_020936076.1 | 725 | 67 | |||
| XM_021080418.1 | XP_020936077.1 | 725 | 67 | |||
| GenBank mRNA | AJ564972.1 | - | 2 | 525 | 67 | |
| AJ564973.1 | - | 2 | 247 | 67 | ||
| AJ564974.1 | - | 3 | 480 | 67 | ||
| AJ564975.1 | - | 3 | 422 | 67 | ||
| AK390511.1 | - | 2 | 408 | 67 | ||
More Information
Set Membership: |
|

PCDH11X
Genomic Map