Assay Details
| Gene Symbol: B3GAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353828.4 | XP_003353876.1 | 440 | 108 | ||
| XM_013994208.2 | XP_013849662.1 | 1000 | 108 | |||
| GenBank mRNA | AJ567374.1 | - | 2 | 145 | 108 | |
| AK233894.1 | - | 2 | 191 | 108 | ||
More Information
Set Membership: |
|

B3GAT3
Genomic Map