Assay Details
| Gene Symbol: POFUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129952.1 | NP_001123424.1 | 1 | 66 | 96 | |
| XM_021077305.1 | XP_020932964.1 | 177 | 96 | |||
| XM_021077306.1 | XP_020932965.1 | 177 | 96 | |||
| GenBank mRNA | AJ567917.1 | - | 1 | 45 | 96 | |
| DQ139939.1 | - | 1 | 66 | 96 | ||
More Information
Set Membership: |
|

POFUT1
Genomic Map