Assay Details
| Gene Symbol: MAPK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354171.5 | XP_003354219.1 | 1031 | 103 | ||
| XM_005661439.3 | XP_005661496.1 | 1001 | 103 | |||
| XM_005661440.3 | XP_005661497.1 | 1102 | 103 | |||
| XM_005661441.3 | XP_005661498.1 | 1035 | 103 | |||
| XM_005661444.3 | XP_005661501.1 | 1200 | 103 | |||
| XM_013994862.2 | XP_013850316.1 | 1032 | 103 | |||
| XM_021084316.1 | XP_020939975.1 | 1084 | 103 | |||
| XM_021084317.1 | XP_020939976.1 | 916 | 103 | |||
| GenBank mRNA | AJ583708.1 | - | 6 | 654 | 103 | |
More Information
Set Membership: |
|

MAPK9
Genomic Map