Assay Details
| Gene Symbol: CXCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135179.5 | XP_003135227.1 | 1294 | 66 | ||
| GenBank mRNA | AJ851240.1 | - | 1 | 456 | 66 | |
| AK237352.1 | - | 2 | 988 | 66 | ||
More Information
Set Membership: |
|

CXCR3
Genomic Map