Assay Details
| Gene Symbol: LOC100515345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075348.1 | XP_020931007.1 | 207 | 148 | ||
| GenBank mRNA | AK237564.1 | - | 2 | 147 | 148 | |
More Information
Set Membership: |
|

LOC100515345
Genomic Map