Assay Details
| Gene Symbol: TFF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358973.3 | XP_003359021.1 | 244 | 76 | ||
| GenBank mRNA | AM283538.1 | - | 2 | 157 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TFF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358973.3 | XP_003359021.1 | 244 | 76 | ||
| GenBank mRNA | AM283538.1 | - | 2 | 157 | 76 | |
Set Membership: |
|