Assay Details
| Gene Symbol: PRKCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066740.1 | XP_020922399.1 | 70 | |||
| GenBank mRNA | AK397783.1 | - | 7 | 716 | 70 | |
| AY093442.1 | - | 7 | 754 | 70 | ||
More Information
Set Membership: |
|

PRKCA
Genomic Map