Assay Details
| Gene Symbol: PRKCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086459.1 | XP_020942118.1 | 8775 | 104 | ||
| XM_021086460.1 | XP_020942119.1 | 2476 | 104 | |||
| GenBank mRNA | AY093443.1 | - | 4 | 354 | 104 | |
More Information
Set Membership: |
|

PRKCB
Genomic Map