Assay Details
| Gene Symbol: SSTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167628.1 | NP_001161100.1 | 1 | 1145 | 112 | |
| XM_005663799.3 | XP_005663856.1 | 1841 | 112 | |||
| XM_005663800.3 | XP_005663857.1 | 1310 | 112 | |||
| GenBank mRNA | AY156054.2 | - | 1 | 1145 | 112 | |
More Information
Set Membership: |
|

SSTR3
Genomic Map