Assay Details
| Gene Symbol: IRF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252427.2 | NP_001239356.1 | 8 | 1124 | 66 | |
| XM_021093479.1 | XP_020949138.1 | 2438 | 66 | |||
| XM_021093480.1 | XP_020949139.1 | 1291 | 66 | |||
| GenBank mRNA | AY435218.2 | - | 8 | 1124 | 66 | |
More Information
Set Membership: |
|

IRF8
Genomic Map