Assay Details
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY625258.1 | - | 1 | 249 | 79 | |
| FJ887980.1 | - | 1 | 532 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY625258.1 | - | 1 | 249 | 79 | |
| FJ887980.1 | - | 1 | 532 | 79 | ||
Set Membership: |
|