Assay Details
| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095458.1 | XP_020951117.1 | 397 | 80 | ||
| XM_021095459.1 | XP_020951118.1 | 397 | 80 | |||
| XM_021095460.1 | XP_020951119.1 | 997 | 80 | |||
| GenBank mRNA | AY653213.1 | - | 1 | 151 | 80 | |
| KX815346.1 | - | 2 | 268 | 80 | ||
More Information
Set Membership: |
|

PAX7
Genomic Map