Assay Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077470.1 | XP_020933129.1 | 1006 | 60 | ||
| GenBank mRNA | AY820766.1 | - | 1 | 54 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A4RG| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077470.1 | XP_020933129.1 | 1006 | 60 | ||
| GenBank mRNA | AY820766.1 | - | 1 | 54 | 60 | |
Set Membership: |
|