Assay Details
| Gene Symbol: GYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195508.1 | NP_001182437.1 | 16 | 2203 | 101 | |
| GenBank mRNA | AK389223.1 | - | 3 | 390 | 101 | |
| AY870324.1 | - | 11 | 1151 | 101 | ||
| GQ845034.1 | - | 16 | 2203 | 101 | ||
More Information
Set Membership: |
|

GYS1
Genomic Map