Assay Details
| Gene Symbol: NR1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067026.1 | XP_020922685.1 | 2074 | 61 | ||
| XM_021067027.1 | XP_020922686.1 | 758 | 61 | |||
| XM_021067028.1 | XP_020922687.1 | 754 | 61 | |||
| GenBank mRNA | DQ120776.1 | - | 2 | 154 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021067028.1 transcripts.
Set Membership: |
|

NR1D1
Genomic Map