Assay Details
| Gene Symbol: S100A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090794.1 | XP_020946453.1 | 127 | 81 | ||
| GenBank mRNA | EF055892.1 | - | 1 | 62 | 81 | |
| Gene Symbol: LOC110260465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090793.1 | XP_020946452.1 | 127 | 81 | ||
More Information
Set Membership: |
|

S100A7
Genomic Map