Assay Details
| Gene Symbol: IRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253352.1 | NP_001240281.1 | 1 | 181 | 87 | |
| XM_021097964.1 | XP_020953623.1 | 274 | 87 | |||
| XM_021097965.1 | XP_020953624.1 | 1354 | 87 | |||
| XM_021097966.1 | XP_020953625.1 | 335 | 87 | |||
| XM_021097967.1 | XP_020953626.1 | 274 | 87 | |||
| XM_021097968.1 | XP_020953627.1 | 335 | 87 | |||
| XM_021097969.1 | XP_020953628.1 | 274 | 87 | |||
| XM_021097970.1 | XP_020953629.1 | 335 | 87 | |||
| GenBank mRNA | EF055893.2 | - | 1 | 181 | 87 | |
More Information
Set Membership: |
|

IRF4
Genomic Map