Assay Details
| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162402.1 | NP_001155874.1 | 1 | 586 | 86 | |
| XM_005656117.2 | XP_005656174.1 | 764 | 86 | |||
| GenBank mRNA | AK395783.1 | - | 2 | 758 | 86 | |
| EF137953.1 | - | 1 | 347 | 86 | ||
| EF546800.1 | - | 1 | 586 | 86 | ||
More Information
Set Membership: |
|

LPAR3
Genomic Map