Assay Details
| Gene Symbol: CCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256774.1 | NP_001243703.1 | 2 | 301 | 73 | |
| GenBank mRNA | AK394989.1 | - | 2 | 332 | 73 | |
| AY934805.2 | - | 2 | 212 | 73 | ||
| EF179159.1 | - | 2 | 153 | 73 | ||
| FJ156753.1 | - | 2 | 301 | 73 | ||
More Information
Set Membership: |
|

CCL11
Genomic Map