Assay Details
| Gene Symbol: CERS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167631.1 | NP_001161103.1 | 8 | 828 | 105 | |
| XM_005661256.3 | XP_005661313.1 | 1125 | 105 | |||
| GenBank mRNA | AB529856.1 | - | 8 | 828 | 105 | |
| AK233295.1 | - | 9 | 911 | 105 | ||
| AK345947.1 | - | 9 | 911 | 105 | ||
| AK396912.1 | - | 9 | 1002 | 105 | ||
| EU275148.1 | - | 6 | 551 | 105 | ||
More Information
Set Membership: |
|

CERS4
Genomic Map