Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001264.1 | NP_001001264.1 | 10 | 1553 | 81 | |
| XM_013994456.2 | XP_013849910.1 | 1964 | 81 | |||
| XM_021081114.1 | XP_020936773.1 | 2488 | 81 | |||
| XM_021081119.1 | XP_020936778.1 | 2446 | 81 | |||
| GenBank mRNA | AB010969.1 | - | 10 | 1553 | 81 | |
More Information
Set Membership: |
|

WT1
Genomic Map