Assay Details
| Gene Symbol: CCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001532.3 | NP_001001532.1 | 3 | 1061 | 67 | |
| GenBank mRNA | AB090356.2 | - | 3 | 1057 | 67 | |
| AB116555.1 | - | 2 | 1009 | 67 | ||
| AK239023.1 | - | 3 | 1064 | 67 | ||
More Information
Set Membership: |
|

CCR7
Genomic Map