Assay Details
| Gene Symbol: CCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001617.1 | NP_001001617.1 | 3 | 1147 | 71 | |
| XM_013981563.2 | XP_013837017.2 | 1075 | 71 | |||
| XM_013981564.2 | XP_013837018.2 | 1075 | 71 | |||
| XM_013981567.2 | XP_013837021.1 | 1078 | 71 | |||
| XM_021068577.1 | XP_020924236.1 | 1075 | 71 | |||
| GenBank mRNA | AB119273.1 | - | 3 | 1147 | 71 | |
| AB119274.1 | - | 2 | 1028 | 71 | ||
| AK348994.1 | - | 2 | 1212 | 71 | ||
More Information
Set Membership: |
|

CCRL2
Genomic Map