Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001642.2 | NP_001001642.1 | 5 - 6 | 351 | 94 | |
| XM_005655543.3 | XP_005655600.1 | 391 | 94 | |||
| XM_021091200.1 | XP_020946859.1 | 489 | 94 | |||
| XM_021091201.1 | XP_020946860.1 | 590 | 94 | |||
| GenBank mRNA | AB530143.1 | - | 4 - 5 | 311 | 94 | |
| AK233656.1 | - | 5 - 6 | 337 | 94 | ||
| AK394575.1 | - | 5 - 6 | 343 | 94 | ||
| AY260940.1 | - | 5 - 6 | 353 | 94 | ||
More Information
Set Membership: |
|

PRKAG1
Genomic Map