Assay Details
| Gene Symbol: CCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001866.1 | NP_001001866.1 | 7 | 649 | 92 | |
| GenBank mRNA | AY573056.1 | - | 7 | 649 | 92 | |
| AY610179.1 | - | 7 | 707 | 92 | ||
More Information
Set Membership: |
|

CCS
Genomic Map