Assay Details
| Gene Symbol: PRLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001868.1 | NP_001001868.1 | 8 | 1755 | 63 | |
| GenBank mRNA | AY308824.1 | - | 8 | 1755 | 63 | |
| AY308825.1 | - | 8 | 1755 | 63 | ||
| AY308826.1 | - | 8 | 1755 | 63 | ||
| DQ157757.1 | - | 8 | 1800 | 63 | ||
| HM059030.1 | - | 1 | 900 | 63 | ||
| HM059031.1 | - | 1 | 900 | 63 | ||
| HM059032.1 | - | 1 | 900 | 63 | ||
| HM059033.1 | - | 1 | 900 | 63 | ||
| HM209402.1 | - | 8 | 1795 | 63 | ||
More Information
Set Membership: |
|

PRLR
Genomic Map