Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001911.1 | NP_001001911.1 | 5 | 753 | 66 | |
| GenBank mRNA | AK349951.1 | - | 5 | 1086 | 66 | |
| AY290706.1 | - | 5 | 753 | 66 | ||
More Information
Set Membership: |
|

KLK1
Genomic Map