Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003662.1 | NP_001003662.1 | 5 - 6 | 950 | 64 | |
| GenBank mRNA | AJ715530.1 | - | 5 - 6 | 950 | 64 | |
| AJ783901.1 | - | 5 - 6 | 950 | 64 | ||
| AK347421.1 | - | 5 - 6 | 1512 | 64 | ||
| AK348272.1 | - | 5 - 6 | 1333 | 64 | ||
| AK395289.1 | - | 6 - 7 | 1172 | 64 | ||
More Information
Set Membership: |
|

FST
Genomic Map