Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003924.1 | NP_001003924.1 | 3 | 347 | 57 | |
| XM_021093596.1 | XP_020949255.1 | 904 | 57 | |||
| GenBank mRNA | AY349424.1 | - | 3 | 347 | 57 | |
More Information
Set Membership: |
|

C1QA
Genomic Map