Assay Details
| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004026.1 | NP_001004026.1 | 4 | 546 | 85 | |
| GenBank mRNA | X51977.1 | - | 546 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004026.1 | NP_001004026.1 | 4 | 546 | 85 | |
| GenBank mRNA | X51977.1 | - | 546 | 85 | ||
Set Membership: |
|