Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004034.1 | NP_001004034.1 | 3 | 455 | 77 | |
| GenBank mRNA | M21683.1 | - | 3 | 455 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004034.1 | NP_001004034.1 | 3 | 455 | 77 | |
| GenBank mRNA | M21683.1 | - | 3 | 455 | 77 | |
Set Membership: |
|