Assay Details
| Gene Symbol: CA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004038.1 | NP_001004038.1 | 8 | 954 | 130 | |
| XM_005664669.3 | XP_005664726.1 | 1658 | 130 | |||
| XM_013998572.2 | XP_013854026.1 | 1447 | 130 | |||
| GenBank mRNA | AY075024.1 | - | 8 | 954 | 130 | |
More Information
Set Membership: |
|

CA11
Genomic Map