Assay Details
| Gene Symbol: TMBIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005348.1 | NP_001005348.1 | 9 | 749 | 151 | |
| GenBank mRNA | AY550048.1 | - | 7 | 506 | 151 | |
| AY713975.1 | - | 9 | 749 | 151 | ||
More Information
Set Membership: |
|

TMBIM6
Genomic Map