Assay Details
| Gene Symbol: KCNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006593.2 | NP_001006593.1 | 1 | 1957 | 108 | |
| GenBank mRNA | AY635585.1 | - | 1 | 1684 | 108 | |
| DQ005442.1 | - | 1 | 1957 | 108 | ||
More Information
Set Membership: |
|

KCNA5
Genomic Map