Assay Details
| Gene Symbol: TAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007196.1 | NP_001007197.1 | 4 - 5 | 353 | 71 | |
| XM_005663900.3 | XP_005663957.2 | 3999 | 71 | |||
| GenBank mRNA | AY769936.1 | - | 4 - 5 | 353 | 71 | |
More Information
Set Membership: |
|

TAC3
Genomic Map