Assay Details
| Gene Symbol: NPPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008482.1 | NP_001008482.1 | 2 | 252 | 135 | |
| XM_005672279.2 | XP_005672336.1 | 415 | 135 | |||
| GenBank mRNA | AK352468.1 | - | 2 | 400 | 135 | |
More Information
Set Membership: |
|

NPPC
Genomic Map